EMAIL THIS PAGE TO A FRIEND
The Canadian veterinary journal = La revue veterinaire canadienne
Antiemetic efficacy of promethazine on xylazine-induced emesis in cats.
PMID 22851784
Abstract
The prophylactic antiemetic effect of 3 dosages of promethazine injected into cats 1 h before administration of xylazine was compared with that of a saline solution. Prior treatment with 2 and 4 mg/kg of promethazine significantly reduced the frequency of emetic episodes. Promethazine may be used as a prophylactic antiemetic in cats treated with xylazine.