EMAIL THIS PAGE TO A FRIEND
Journal of food science
Stimulation of cadaverine production by foodborne pathogens in the presence of Lactobacillus, Lactococcus, and Streptococcus spp.
PMID 22853653
Abstract
The effect of Lactobacillus plantarum (FI8595), Lactococcus lactis subsp. cremoris MG 1363), Lactococcus lactis subsp. lactis (IL 1403), and Streptococcus thermophilus on cadaverine and other biogenic amine production by foodborne pathogens was investigated lysine decarboxylase broth. Both of lactic acid bacteria and foodborne pathogens used (especially Staphylococcus aureus, E. coli, Lc. lactis subsp. lactis and Lb. plantarum) had an ability to convert aminoacids into biogenic amine. The conversion of lysine into cadaverine was the highest (167.11 mg/L) by Lactobacillus spp. Gram-positive bacteria generally had a greater ability to produce cadaverine with corresponding value of 46.26, 53.76, and 154.54 mg/L for Enterococcus faecalis, S. aureus, and Listeria monocytogenes, respectively. Significant variations on biogenic amine production were observed in the presence of lactic acid bacteria strains (P < 0.05). The role of lactic acid bacteria on biogenic amine production by foodborne pathogens varied depending on strains and specific amine. Cadaverine accumulation by Enterobactericeae was increased in the presence of lactic acid bacteria strains except for St. thermophilus, which induced 2-fold lower cadaverine production by S. Paratyphi A. Lc. lactis subsp. lactis and Lc. lactis subsp. cremoris induced 10-fold higher increases in histamine for E. coli and K. pneumoniae, respectively. Lactic acid bacteria resulted in strong increases in cadaverine production by P. aeruginosa, although remarkable decreases were observed for histamine, spermidine, dopamine, agmatine, and TMA in the presence of lactic acid bacteria in lysine decarboxylase broth . The result of the study showed that amine positive lactic acid bacteria strains in fermented food led to significant amine accumulation by contaminant bacteria and their accumulation in food product may be controlled by the use of proper starters with amine-negative activity.