EMAIL THIS PAGE TO A FRIEND
Organic letters
Short synthesis of diamide-linked sucrose macrocycles.
PMID 22853688
Abstract
A convenient route to macrocyclic diamide-linked macrocyclic derivatives with a sucrose scaffold is presented. Reaction of sucrose based amines (o- and m-) with acid dichlorides afforded the monomeric macrocycles in excellent yields, while reaction of the p-amines also provided dimeric products.