EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
The synthesis of 2-deoxy-α-D-glycosides from D-glycals catalyzed by TMSI and PPh3.
PMID 22854184
Abstract
2-Deoxyglycosides were synthesized in high α-selectivity by the direct addition of alcohols to D-glucal and D-galactal catalyzed by TMSI and PPh(3). The acid labile isopropylidene group is tolerated under this condition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
464058
D-Glucal, 96%
C6H10O4