EMAIL THIS PAGE TO A FRIEND
Zhongguo ying yong sheng li xue za zhi = Zhongguo yingyong shenglixue zazhi = Chinese journal of applied physiology
[Application of recording SK2 current in human atrial myocytes by perforated patch clamp techniques with the mix of beta-escin and amphotericin B].
PMID 22860418
Abstract
To establish a perforated patch-clamp technology with amphotericin B and beta-escin and to research the regulation of small conductance calcium-activated potassium channel SK2 currents by calcium ions. Single human atrial myocytes were enzymatically isolated from the right atrial appendage. Amphotericin B and / or beta-escin were used by perforated electrode liquid. The regulation of SK2 current by calcium ions in human atrial myocytes was performed with the perforated patch-clamp technique. The intracellular calcium changes were measured by the intracellular calcium test system. Mixed perforated electrode liquid compared with 150 microg/ml amphotericin B or 6.88 microg/ml beta-escin alone, it was easy to seal cells and activate SK2 current by the former method. Moreover, the ration of F340/380 was consistent with the change of intracellular free calcium ion concentration increase after the formation of perforation. The ration of F340/380 was measured by intracellular calcium test system. The appropriate concentration of amphotericin B mixed with beta-escin can form a stable whole-cell patch recording technology that is appropriate for the research of SK2 current regulation by intracellular calcium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1378 Escin, ≥95%, powder