EMAIL THIS PAGE TO A FRIEND
European journal of internal medicine
Lymphocyte-suppressing effects of omega-3 fatty acids in bezafibrate-treated patients with isolated hypertriglyceridemia.
PMID 22863445