EMAIL THIS PAGE TO A FRIEND
Dermatology online journal
Letter: Tinea capitis in infants in their first 2 years of life: A 12-year study and a review of the literature.
PMID 22863638
Abstract
Tinea capitis (TC) is a dermatophyte infection that occurs mainly in childhood; but it is uncommon in infants. The aim of this study was to review the clinical and mycological profile of TC in infants. We present a retrospective study; we enrolled all the cases of infant TC over a period of 12 years (1999-2010). Thirty-five infants (21 boys, 14 girls) with a mean age of 20.16 months were diagnosed with TC among a total number of 881 cases of TC (3.9%). Scalp scaling and alopecia were the most frequent clinical features. Microsporic tinea (21 cases) was the most frequent followed by Trichophytic tinea (9 cases) and inflammatory tinea (5 cases). Direct microscopy of hair was positive in 33 cases (94.2%). Culture positivity was found in 82.8 percent of infants (29 cases). Four species of dermatophytes were isolated; Microsporum canis in 18 cases (62%) followed by Trichophyton violaceum, Trichophyton mentagrophytes, and Trichophyton verrucosum. Twenty-nine infants were treated successfully with griseofulvin. TC is rare in infants. The diagnosis of TC should be considered if scaling and/or alopecia are present and should be confirmed by mycology testing prior to initiation of treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4753
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%
C17H17ClO6