EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Amphibacillus marinus sp. nov., a member of the genus Amphibacillus isolated from marine mud.
PMID 22863987
Abstract
A gram-positive, spore-forming, rod-shaped bacterium, designated J1(T) was isolated from deep-sea mud collected from the South China Sea and subjected to polyphasic taxonomic investigation. Phylogenetic analysis based on 16S rRNA gene sequences revealed that J1(T) clustered with the type strains of Amphibacillus cookii, Amphibacillus sediminis and Amphibacillus jilinensis and exhibited a range of similarity of 93.9-97.0 % to members of the genus Amphibacillus. The DNA G+C content was 36.7 mol%. Chemotaxonomic analysis showed no quinones, and the cell wall contained meso-diaminopimelic acid as the diagnostic diamino acid for strain J1(T). The major cellular fatty acids were iso-C15 : 0 and anteiso-C15 : 0. Strain J1(T) was positive for catalase activity and negative for oxidase activity. On the basis of phylogenetic position and phenotypic properties, strain J1(T) represents a novel species of the genus Amphibacillus and the name Amphibacillus marinus sp. nov. is proposed. The type strain is J1(T) ( = CGMCC 1.10434(T) = JCM 17099(T)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33240
2,6-Diaminopimelic acid, ≥97.0% (NT)
C7H14N2O4
D1377
2,6-Diaminopimelic acid, ≥98% (TLC)
C7H14N2O4