EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Extraction of betulin, trimyristin, eugenol and carnosic acid using water-organic solvent mixtures.
PMID 22864237
Abstract
A solvent system consisting of ethyl acetate, ethyl alcohol and water, in the volume ratio of 4.5:4.5:1, was developed and used to extract, at room temperature, betulin from white birch bark and antioxidants from spices (rosemary, thyme, sage, and oregano) and white oak chips. In addition, under reflux conditions, trimyristin was extracted from nutmeg using the same solvent system, and eugenol from olives was extracted using a mixture of salt water and ethyl acetate. The protocol demonstrates the use of water in organic solvents to extract natural products from plants. Measurement of the free-radical scavenging activity using by 2,2-diphenyl-1-picrylhydrazyl (DPPH) indicated that the extraction of plant material using ethyl acetate, ethyl alcohol and water (4.5:4.5:1, v/v/v) was exhaustive when carried out at room temperature for 96 h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9757
Betulin, ≥98%
C30H50O2
92648
Betulin, analytical standard
C30H50O2
T5141
Glyceryl trimyristate, ≥99%
C45H86O6