EMAIL THIS PAGE TO A FRIEND
Molecules (Basel)
A convenient ultrasound-promoted synthesis of some new thiazole derivatives bearing a coumarin nucleus and their cytotoxic activity.
PMID 22864241
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34857
1,4-Dioxane, for HPLC, ≥99.5%
C4H8O2
360481
1,4-Dioxane, ACS reagent, ≥99.0%
C4H8O2
676934
1,4-Dioxane, ACS reagent, ≥99.0%
C4H8O2
33147
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C4H8O2
D201863
1,4-Dioxane, ReagentPlus®, ≥99%
C4H8O2
CRM48367
1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL
C4H8O2
612073
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.
C4H8O2
611905
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.
C4H8O2
551368
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D)
C4H8O2
612111
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
631833
1,4-Dioxane solution, NMR reference standard, 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
631868
1,4-Dioxane solution, NMR reference standard, 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
684228
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
C4H8O2
704806
1,4-Dioxane solution, NMR reference standard, 1 mM in D2O ("100", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
704814
1,4-Dioxane solution, NMR reference standard, 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
704822
1,4-Dioxane solution, NMR reference standard, 0.1 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
736171
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.
C4H8O2
703354
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 6.5 mm × 8 in.
C4H8O2
77450
2-Bromoacetophenone, for GC derivatization, ≥99.0%
C8H7BrO
115835
2-Bromoacetophenone, 98%
C8H7BrO
10905
Chloroacetone, produced by Wacker Chemie AG, Burghausen, Germany, ≥96.0% (GC)
C3H5ClO
167479
Chloroacetone, 95%
C3H5ClO