EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Total synthesis of ascididemin via anionic cascade ring closure.
PMID 22864261
Abstract
A new and convergent synthesis of ascididemin is presented. Using an anionic cascade ring closure as the key step, this natural product is obtained in 45% overall yield in just 6 steps starting from 2'-fluoroacetophenone. This new approach was extended to the synthesis of a new isomer of ascididemin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183717
2′-Fluoroacetophenone, 97%
C8H7FO
548030
3-Bromo-4-methylpyridine, 96%
C6H6BrN