EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
H-point curve isolation method for determination of catechol in complex unknown mixtures.
PMID 22864391
Abstract
In this work, the combination of H-point curve isolation method (HPCIM) and H-point standard additions method (HPSAM) was used for determination of catechol in the presence of phenolic interferents. Spectrophotometric multivariate calibration data constructed by successive standard additions of an analyte in an unknown matrix was used by the method. A cumulative spectrum for interferents in sample was extracted by HPCIM and then HPSAM is used for determination of the catechol concentration by obtained cumulative interferents spectrum. The method was tested with simulated data set. The spectrum obtained from applying HPCIM to the simulated data well agrees with the cumulative spectra of the interferents. The method was applied to the determination of catechol in the presence of highly overlapping interferents in synthetic ternary mixtures using spectrophotometric data. Moreover, the proposed method was successfully used for determination of catechol in real complicated matrices of tea and urine samples. Percent recoveries were between 95.4 and 113.6.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53363
Resorcinol, certified reference material, TraceCERT®
C6H6O2