EMAIL THIS PAGE TO A FRIEND
Arerugi = [Allergy]
[Drug allergy].
PMID 22868514
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1755
Aminophylline, ≥98%, powder
C7H8N4O2 · 0.5C2H8N2