EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section F, Structural biology and crystallization communications
Binding of the unreactive substrate analog L-2-amino-3-guanidinopropionic acid (dinor-L-arginine) to human arginase I.
PMID 22869115
Abstract
Human arginase I (HAI) is a binuclear manganese metalloenzyme that catalyzes the hydrolysis of L-arginine to form L-ornithine and urea through a metal-activated hydroxide mechanism. Since HAI regulates L-Arg bioavailability for NO biosynthesis, it is a potential drug target for the treatment of cardiovascular diseases such as atherosclerosis. X-ray crystal structures are now reported of the complexes of Mn(2)(2+)-HAI and Co(2)(2+)-HAI with L-2-amino-3-guanidinopropionic acid (AGPA; also known as dinor-L-arginine), an amino acid bearing a guanidinium side chain two methylene groups shorter than that of L-arginine. Hydrogen bonds to the α-carboxylate and α-amino groups of AGPA dominate enzyme-inhibitor recognition; the guanidinium group does not interact directly with the metal ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5402
L-2-Amino-3-guanidinopropionic acid hydrochloride
C4H10N4O2 · HCl