PloS one
PRRT2 mutations in paroxysmal kinesigenic dyskinesia with infantile convulsions in a Taiwanese cohort.
PMID 22870186
Abstract
Mutations in the PRRT2 gene have recently been identified in patients with familial paroxysmal kinesigenic dyskinesia with infantile convulsions (PKD/IC) and patients with sporadic PKD/IC from several ethnic groups. To extend these recent genetic reports, we investigated the frequency and identities of PRRT2 mutations in a cohort of Taiwanese patients with PKD/IC. We screened all 3 coding exons of PRRT2 for mutations in 28 Taiwanese patients with PKD/IC. Among them, 13 had familial PKD/IC and 15 were apparently sporadic cases. In total, 7 disparate mutations were identified in 13 patients, including 8 familial cases and 5 apparently sporadic cases. The mutations were not present in 500 healthy controls. Four mutations were novel. One patient had a missense mutation and all other patients carried PRRT2 mutations putatively resulting in a protein truncation. Haplotype analysis revealed that 5 of the 7 patients with the PRRT2 p.R217Pfs*8 mutation shared the same haplotype linked to the mutation. PRRT2 mutations account for 61.5% (8 out of 13) of familial PKD/IC and 33.3% (5 out of 15) of apparently sporadic PKD/IC in the Taiwanese cohort. Most patients with the PRRT2 p.R217Pfs*8 mutation in Taiwan likely descend from a single common ancestor. This study expands the spectrum of PKD/IC-associated PRRT2 mutations, highlights the pathogenic role of PRRT2 mutations in PKD/IC, and suggests genetic heterogeneity within idiopathic PKD.