EMAIL THIS PAGE TO A FRIEND
Future cardiology
Cardiotoxicity in childhood cancer survivors: strategies for prevention and management.
PMID 22871201
Abstract
Advances in cancer treatment have greatly improved survival rates of children with cancer. However, these same chemotherapeutic or radiologic treatments may result in long-term health consequences. Anthracyclines, chemotherapeutic drugs commonly used to treat children with cancer, are known to be cardiotoxic, but the mechanism by which they induce cardiac damage is still not fully understood. A higher cumulative anthracycline dose and a younger age of diagnosis are only a few of the many risk factors that identify the children at increased risk of developing cardiotoxicity. While cardiotoxicity can develop at anytime, starting from treatment initiation and well into adulthood, identifying the best cardioprotective measures to minimize the long-term damage caused by anthracyclines in children is imperative. Dexrazoxane is the only known agent to date, that is associated with less cardiac dysfunction, without reducing the oncologic efficacy of the anthracycline doxorubicin in children. Given the serious long-term health consequences of cancer treatments on survivors of childhood cancers, it is essential to investigate new approaches to improving the safety of cancer treatments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1446
Dexrazoxane, ≥95% (HPLC)
C11H16N4O4
R8657
Razoxane, >98% (HPLC)
C11H16N4O4