EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Fast and reversibly switchable wettability induced by a photothermal effect.
PMID 22871674
Abstract
We report a novel approach for the fabrication of a photo-responsive surface with fast and reversibly switchable wettability between hydrophobicity and hydrophilicity induced by a photothermal effect.