EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Chiral Brønsted acid catalyzed enantioselective allenylation of aldehydes.
PMID 22872133
Abstract
A versatile and highly enantioselective chiral Brønsted acid-catalyzed allenylation of aldehydes with propargyl borolane is reported. The reaction is shown to be practical and quite general with a broad substrate scope covering aryl, heteroaryl, α,β-unsaturated, and aliphatic aldehydes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M74253
N-Methyl-N-propargylbenzylamine, 97%
C11H13N
P8013
Pargyline hydrochloride
C11H13N · HCl