EMAIL THIS PAGE TO A FRIEND
Sudebno-meditsinskaia ekspertiza
[Chemical toxicological identification of esfenvalerate].
PMID 22876649
Abstract
The optimal conditions for the isolation of esfenvalerate from the biological specimens have been determined. It was shown that this compound can be separated from the endogenous component of a biological material by means of liquid-liquid extraction and chromatography on a L 40/100 mcm silica gel column. The identification and quantitative analysis of esfenvalerate extracted from cadaveric liver and blood can be accomplished with the use of thin layer chromatography, UV and IR spectrophotometry as well as chromatomass spectrometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
360481
1,4-Dioxane, ACS reagent, ≥99.0%
C4H8O2
34857
1,4-Dioxane, for HPLC, ≥99.5%
C4H8O2
76887
1,4-Dioxane, analytical standard
C4H8O2
D201863
1,4-Dioxane, ReagentPlus®, ≥99%
C4H8O2
33147
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C4H8O2
676934
1,4-Dioxane, ACS reagent, ≥99.0%
C4H8O2
CRM48367
1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL
C4H8O2
736171
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.
C4H8O2
551368
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D)
C4H8O2
611905
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.
C4H8O2
684228
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
C4H8O2
704814
1,4-Dioxane solution, NMR reference standard, 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
612111
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
612073
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.
C4H8O2
631833
1,4-Dioxane solution, NMR reference standard, 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
631868
1,4-Dioxane solution, NMR reference standard, 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
704806
1,4-Dioxane solution, NMR reference standard, 1 mM in D2O ("100", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
704822
1,4-Dioxane solution, NMR reference standard, 0.1 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
703354
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 6.5 mm × 8 in.
C4H8O2
46277
Esfenvalerate, PESTANAL®, analytical standard
C25H22ClNO3
45495
Fenvalerate, PESTANAL®, analytical standard
C25H22ClNO3
F1428
Fenvalerate, ≥97%
C25H22ClNO3