EMAIL THIS PAGE TO A FRIEND
Biomaterials
Direct thrombin inhibitor-bivalirudin functionalized plasma polymerized allylamine coating for improved biocompatibility of vascular devices.
PMID 22877639
Abstract
The direct thrombin inhibitor of bivalirudin (BVLD), a short peptide derived from hirudin, has drawn an increasing attention in clinical application because it is safer and more effective than heparin for diabetic patients with moderate- or high-risk for acute coronary syndromes (ACS). In this study, BVLD was covalently conjugated on plasma polymerized allylamine (PPAam) coated 316L stainless steel (SS) to develop an anticoagulant surface. QCM-D real time monitoring result shows that 565±20 ng/cm2 of BVLD was bound to the PPAam surface. Infrared spectroscopy (IR) and X-ray photoelectron spectroscopy (XPS) confirmed the immobilization of BVLD. The conjugation of BVLD onto the PPAam coating led to enhanced binding of thrombin, and the activity of the thrombin adsorbed on its surface was effectively inhibited. As a result, the BVLD immobilized PPAam (BVLD-PPAam) substrate prolonged the clotting times, and exhibited inhibition in adhesion and activation of platelets and fibrinogen. We also found that the BVLD-PPAam coating significantly enhanced endothelial cell adhesion, proliferation, migration and release of nitric oxide (NO) and secretion of prostaglandin I2 (PGI2). In vivo results indicate that the BVLD-PPAam surface restrained thrombus formation by rapidly growing a homogeneous and intact endothelium on its surface. These data suggest the potential of this multifunctional BVLD-PPAam coating for the application not only in general vascular devices such as catheters, tubes, oxygenator, hemodialysis membranes but also vascular grafts and stents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241075
Allylamine, ≥99%
C3H7N
145831
Allylamine, 98%
C3H7N