EMAIL THIS PAGE TO A FRIEND
Revue belge de medecine dentaire
[Reducing the impact of certain nuisances in the dental office. Working methods and arrangement of the premises].
PMID 2287775
Abstract
The activity of the dentists generates nuisances due, for instance, to the use of biocontaminating aerosols and the emission of waste containing mercury. These two points are thoroughly reviewed in order to raise risk-awareness. As first and main victims of these nuisances, dentists must learn preventive methods and how to implement them. Anti-polluting methods are explained which are effective and easy to apply though sometimes not very well know and IDF recommendations are recalled.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5200
Triparanol, ≥97% (HPLC), powder
C27H32NO2Cl