EMAIL THIS PAGE TO A FRIEND
Journal of hepatology
Patatin-like phospholipase domain-containing 3 (PNPLA3) I148M (rs738409) affects hepatic VLDL secretion in humans and in vitro.
PMID 22878467
Abstract
The robust association between non-alcoholic fatty liver disease (NAFLD) and the genetic variant I148M (rs738409) in PNPLA3 has been widely replicated. The aim of this study was to investigate the effect of the PNPLA3 I148M mutation on: (1) hepatic secretion of very low density lipoproteins (VLDL) in humans; and (2) secretion of apolipoprotein B (apoB) from McA-RH 7777 cells, which secrete VLDL-sized apoB-containing lipoproteins. VLDL kinetics was analyzed after a bolus infusion of stable isotopes in 55 overweight/obese men genotyped for the PNPLA3 I148M variant. Intracellular lipid content, apoB secretion and glycerolipid metabolism were studied in McA-RH 7777 cells overexpressing the human 148I wild type or 148M mutant PNPLA3 protein. In humans, carriers of the PNPLA3 148M allele had increased liver fat compared to 148I homozygotes, and kinetic analysis showed a relatively lower secretion of the large, triglyceride-rich VLDL (VLDL(1)) in 148M carriers vs. 148I homozygotes for the same amount of liver fat. McA-RH 7777 cells overexpressing the 148M mutant protein showed a higher intracellular triglyceride content with a lower apoB secretion and fatty acid efflux, compared to cells overexpressing the 148I wild type protein. The responses with 148M matched those observed in cells expressing the empty vector, indicating that the mutation results in loss of function. We have shown that PNPLA3 affects the secretion of apoB-containing lipoproteins both in humans and in vitro and that the 148M protein is a loss-of-function mutation. We propose that PNPLA3 148M promotes intracellular lipid accumulation in the liver by reducing the lipidation of VLDL.