EMAIL THIS PAGE TO A FRIEND
Nursing children and young people
Developing a nurse-led child sedation service.
PMID 22880310
Abstract
This article explores nurse-led sedation of children in preparation for investigations such as magnetic resonance imaging or minor surgical procedures. The importance of the multidisciplinary team in setting up, implementing and evaluating this new nurse-led initiative is described. Effective planning, involvement, agreement and training are key to a successful change in practice, with responsibility being delegated to competent nursing staff. Evaluation has shown all-round benefits. Trained nurse-led sedation is safe, effective and efficient and nurses are motivated to undertake this extended role.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47335-U
Chloral hydrate solution, 1000 μg/mL in acetonitrile, analytical standard
C2H3Cl3O2