EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
The α7 nicotinic acetylcholine receptor ligands methyllycaconitine, NS6740 and GTS-21 reduce lipopolysaccharide-induced TNF-α release from microglia.
PMID 22884467
Abstract
The anti-inflammatory properties of, particularly the α7, nicotinic acetylcholine receptors (nAChRs) in the peripheral immune system are well documented. There are also reports of anti-inflammatory actions of nicotine in the CNS, but it is unclear, whether this is due to activation or inhibition of nAChRs. Here we investigate the mechanisms behind α7 nAChR-mediated modulation of TNF-α release. We show that α7 nAChR agonists or positive allosteric modulators do not affect LPS-induced release of the pro-inflammatory cytokine TNF-α from cultured microglia. This suggests that classical activation of, i.e. ion-flux through, the α7 nAChR does not reduce TNF-α release from activated microglia. Contrarily, the α7 nAChR antagonist methyllycaconitine and the weak (<10%) agonist NS6740 reduced LPS-induced TNF-α release, indicating that α7 nAChR antagonism conveys anti-inflammatory properties on microglia. The effect of methyllycaconitine or NS6740 was not due to changes in MAPK signaling. These results suggest that the anti-inflammatory effects of nicotine seen in vivo are not due to classical activation of the α7 nAChR, and further suggest that antagonism of α7 nAChRs may reduce neuroinflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0326
GTS-21, ≥97% (HPLC)
C19H20N2O2 · 2HCl