EMAIL THIS PAGE TO A FRIEND
Marine pollution bulletin
Persistent organic pollutants in the endangered Hawaiian monk seal (Monachus schauinslandi) from the main Hawaiian Islands.
PMID 22884537
Abstract
Little is known about levels or effects of persistent organic pollutants (POPs) in Hawaiian monk seals (HMS) from the main Hawaiian Islands (MHI) subpopulation. This study examined concentrations of a large suite of POPs in blubber and serum of juvenile and adult HMS from the MHI. Adult females have the lowest blubber levels of most POPs, whereas adult males have highest levels. POPs in serum were significantly different in adult males compared with adult females for chlordanes and summed dichlorodiphenyltrichloroethanes (DDTs). Lipid-normalized concentrations of chlordanes, DDTs, polychlorinated biphenyls, and mirex in paired blubber and serum samples were significantly correlated. Contaminant levels from the MHI were at similar or lower levels than those from remote Northwestern Hawaiian Island populations. Determining initial ranges of POPs is an important step towards assessing one of the many potential health threats to this critically endangered species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45378
Chlordane (technical mixture), PESTANAL®, analytical standard
C10H6Cl8