EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Optimization of solid phase microextraction for non-lethal in vivo determination of selected pharmaceuticals in fish muscle using liquid chromatography-mass spectrometry.
PMID 22885039
Abstract
A new thin film microextraction (TFME) configuration based on C(18) thin film was optimized to improve solid phase microextraction (SPME) sensitivity and extraction kinetics for in vivo determinations of trace pharmaceuticals in fish tissue. Optimization of SPME involved increasing sampler surface area and volume of extraction phase to improve performance within in vitro applications such as agarose gel matrix and spiked fish tissue, as well as in vivo determinations of pharmaceuticals in fish exposed to municipal wastewater. Rainbow trout (Oncorhynchus mykiss) were exposed for 4d to effluents from a municipal wastewater plant that was treated under three different configurations of a pilot plant. Fathead minnow (Pimephales promelas) were caged for 14 d upstream and downstream of municipal wastewater effluent discharges along Grand River at near Kitchener, ON. TFME and regular C(18) fibers were inserted into the dorsal-epaxial muscle of rainbow trout and fathead minnow, respectively, for 30 min sampling intervals. Sample extracts obtained from fibers were desorbed in methanol:water (3:2) for 90 min under 1500 rpm agitation and analyzed using liquid chromatography coupled with tandem mass spectrometry (LC/MS-MS) to determine pharmaceutical bioconcentration. Most target pharmaceuticals were detected in the wastewater with the exception of norfluoxetine and paroxetine. C(18) TFME phase successfully quantified fluoxetine, venlafaxine, sertraline, paroxetine, and carbamazapine in muscle of living fish at concentrations ranging from 1.7 to 259 ng/g. Reproducibility of the method in spiked fish muscle was 9-18% RSD with limits of detection and quantification ranging from 0.08 to 0.21 ng/g and 0.09 to 0.64 ng/g (respectively) for the analytes examined. Ibuprofen and gemfibrozil were not detected in fish tissues, likely due to rapid excretion leading to low rates of bioconcentration. This study demonstrates improved in vivo SPME sensitivity (recovery) and extraction rates during pre-equilibrium sampling of target pharmaceuticals in fish tissue due to the improved C(18) extraction phase geometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z129178
Aldrich® flask-type sprayer, size 250 mL