EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Ectoine biosynthesis in Mycobacterium smegmatis.
PMID 22885758
Abstract
Mycobacterium smegmatis is a commonly used mycobacterial model system. Here, we show that M. smegmatis protects itself against elevated salinity by synthesizing ectoine and hydroxyectoine and characterize the phenotype of a nonproducing mutant. This is the first analysis of M. smegmatis halotolerance and of the molecular mechanism that supports it.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
81619
Ectoine, ≥95.0% (HPLC)
C6H10N2O2
E2271
Ectoine, osmoprotectant
C6H10N2O2