EMAIL THIS PAGE TO A FRIEND
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians
Cerclage for the prevention of preterm birth in high risk women receiving intramuscular 17-α-hydroxyprogesterone caproate.
PMID 22889234
Abstract
To assess cerclage benefit in women with short cervix also receiving 17-α-hydroxyprogesterone caproate (17P) to prevent recurrent preterm birth (PTB). Secondary analysis of a multicenter trial of ultrasound-indicated cerclage for shortened cervical length (CL). Women with prior spontaneous PTB at 16-33 6/7 weeks, singleton gestation and CL < 25 mm between 16 and 22 6/7 weeks were counseled on use of 17P and randomized to cerclage or no cerclage. Outcomes of women who received 17P were analyzed by randomization group. Primary outcome was PTB < 35 weeks. 99 women received 17P: 47 cerclage; 52 no cerclage. Rates of PTB < 35 weeks were similar, 30% for cerclage and 38% for no cerclage (aOR 0.64 (0.27-1.52)). In women with CL < 15 mm, PTB < 35 weeks was reduced for the cerclage group (17% vs. 75%, p = 0.02). However, this difference was nullified after controlling for total progesterone doses received (p = 0.40). Cerclage was shown not to offer additional benefit for the prevention of recurrent PTB in women with short CL < 25 mm receiving 17P, but the sample size is insufficient for a definite conclusion given the 36% nonsignificant decrease in the odds of PTB < 35 weeks. Cerclage may further offer substantial benefit to women with very short CL < 15 mm and further study is needed.