EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Diagnosis of triclabendazole resistance in Fasciola hepatica.
PMID 22890399
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32802
Triclabendazole, VETRANAL®, analytical standard
C14H9Cl3N2OS