EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Drastic enhancement of excess electron-transfer efficiency through DNA by inserting consecutive 5-phenylethynyl-2'-deoxyuridines as a modulator.
PMID 22890576
Abstract
5-(Phenylethynyl)-2'-deoxyuridine ((Ph)U) was consecutively incorporated into oligodeoxynucleotides as an electron transport modulator for regulating DNA-mediated excess electron transfer (EET). Upon photoexcitation of the ethynylpyrene chromophore, extremely high EET efficiencies through DNA containing consecutive (Ph)Us were observed using 5-bromouracil as a distal electron trap.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5412
2′-Deoxyuridine, ≥98.5%
C9H12N2O5