EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Hair regrowth in a male patient with extensive androgenetic alopecia on estrogen therapy.
PMID 22890743
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2300000
Minoxidil, European Pharmacopoeia (EP) Reference Standard
C9H15N5O