EMAIL THIS PAGE TO A FRIEND
Journal of dietary supplements
An evidence-based systematic review of vanadium by the Natural Standard Research Collaboration.
PMID 22891992
Abstract
An evidence-based systematic review of vanadium by the Natural Standard Research Collaboration consolidates the safety and efficacy data available in the scientific literature using a validated, reproducible grading rationale. This article includes written and statistical analysis of clinical trials, plus a compilation of expert opinion, folkloric precedent, history, pharmacology, kinetics/dynamics, interactions, adverse effects, toxicology, and dosing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357162
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
V
357170
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
V
266205
Vanadium, foil, thickness 0.5 mm, 99.7% trace metals basis
V
262935
Vanadium, powder, −325 mesh, 99.5% trace metals basis
V
266175
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
V
262927
Vanadium, turnings, 99.7% trace metals basis
V
774073
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
V
774065
Vanadium, powder, -100 mesh, 99.9% trace metals basis
V
GF98785388
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF25800386
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
V
GF46548692
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF21697106
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF76957004
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF23355996
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF69763977
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
V
GF75438445
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
V
GF84027269
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
V
GF25047503
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF39723239
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
V
GF29805341
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
V
GF03408555
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
V
GF30937732
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
V
GF57656391
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
V
GF25668344
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
V
GF26068855
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
V
GF67217430
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
V
GF78288946
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
V
GF08542975
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
V
GF58606409
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
V
GF02179672
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
V