EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Absorption characteristics of new solvent based on a blend of AMP and 1,8-diamino-p-menthane for CO2 absorption.
PMID 22893968
Abstract
Aqueous 1,8-diamino-p-menthane (KIER-C3) and commercially available amine solutions were tested for CO2 absorption. A 2-amino-2-methyl-1-propanol (AMP) solution with an addition of KIER-C3 showed 9.3% and 31.6% higher absorption rate for CO2 than the AMP solution with an addition of monoethanolamine (MEA) and ammonia (NH3), respectively. The reaction rate constant for CO2 absorption by the AMP/KIER-C3 solution was determined by the following equation: k2,AMP/C3 = 7.702 x 10(6) exp (-2248.03/T). A CO2 loading ratio of the AMP/KIER-C3 solution was also 2 and 3.4-times higher than that of the AMP/NH3 solution and the AMP/MEA solution, respectively. Based on the experimental results, KIER-C3 may be used as an excellent additive to increase CO2 absorption capability of AMP.