EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Copper-catalyzed oxidative amidation of aldehydes with amine salts: synthesis of primary, secondary, and tertiary amides.
PMID 22894712
Abstract
A practical method for the amidation of aldehydes with economic ammonium chloride or amine hydrochloride salts has been developed for the synthesis of a wide variety of amides by using inexpensive copper sulfate or copper(I) oxide as a catalyst and aqueous tert-butyl hydroperoxide as an oxidant. This amidation reaction is operationally straightforward and provides primary, secondary, and tertiary amides in good to excellent yields for most cases utilizing inexpensive and readily available reagents under mild conditions. In situ formation of amine salts from free amines extends the substrate scope of the reaction. Chiral amides are also synthesized from their corresponding chiral amines without detectable racemization. The practicality of this amide formation reaction has been demonstrated in an efficient synthesis of the antiarrhythmic drug N-acetylprocainamide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
269476
N-Acetylprocainamide, ≥99%
C15H23N3O2
A5513
N-Acetylprocainamide hydrochloride, ≥99% (HPLC), powder
C15H23N3O2 · HCl