EMAIL THIS PAGE TO A FRIEND
BMC research notes
Analysis of naltrexone and its metabolite 6-beta-naltrexol in serum with high-performance liquid chromatography.
PMID 22894733
Abstract
Naltrexone has been proven to be an effective treatment option for the treatment of alcohol dependency. In this article we introduce a reliable and simple method developed for the simultaneous determination of naltrexone and 6-β-naltrexol in human serum by using high-performance liquid chromatography (HPLC). Liquid-liquid extraction with butyl acetate from basic solutions (pH 9) was chosen for extraction with nalorphine as an internal standard (IS). Analytes were back-extracted from organic solvent into perchloric acid. The acid extract was chromatographed by HPLC with a reverse-phase ODS-column and electrochemical detector. The mobile phase was a NaH(2)PO(4)-solution with acetonitrile as an organic modifier and octanesulphonic acid and tetraethylammonium hydrogen sulphate as ion-pair reagents. The recovery of the extraction method was 48% for naltrexone and 75% for 6-β-naltrexol. The limit of quantification was 5.0 ng/ml for naltrexone and 1.0 ng/ml for 6-β-naltrexol. The analysed concentrations of naltrexone differed from the theoretic concentrations by 0.7 to 2.3% and those of 6-β-naltrexol by 2.6%. The relative standard deviation of within-day assay was from 0.9 to 5.7% for naltrexone and from 0.8 to 4.2% for 6-β-naltrexol; for the between-day assay it was 5.7% and 4.2%, respectively. Our results indicate that the developed method is suitable for determination of naltrexone and 6-β-naltrexol in human serum.