EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
Combination treatment with bortezomib and thiostrepton is effective against tumor formation in mouse models of DEN/PB-induced liver carcinogenesis.
PMID 22894930
Abstract
Nanoparticle-encapsulated thiazole antibiotic, thiostrepton, has been shown to be an effective agent for inhibiting tumor growth in solid tumor models through the inhibition of proteasomal activity by the induction of apoptosis in cancer cells. Here, we show the efficacy of thiostrepton-micelles in inhibiting tumor growth in a DEN/PB-induced liver cancer model. We also demonstrate an enhanced anticancer effect of the combination treatment of thiostrepton with bortezomib, another proteasome inhibitor in this liver cancer model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8902
Thiostrepton from Streptomyces azureus, ≥90% (HPLC)
C72H85N19O18S5