EMAIL THIS PAGE TO A FRIEND
Plant signaling & behavior
The role of peroxidases on the mode of action of chalcone in Arabidopsis roots.
PMID 22895105
Abstract
Chalcone is a secondary metabolite belonging to the group of flavonoids. It has shown strong phytotoxic activity on Arabidopsis roots, as inductor of programmed cell death, and inhibitor of root growth and root hair formation. Peroxidases are particularly abundant in root meristems and are involved in the formation and interconversion of reactive oxygen species (ROS), which play a critical role on root and root hair development. Therefore, we report here the role of peroxidases in Arabidopsis root development during chalcone treatment. A strong inhibition of peroxidase activity was detected in the apical root meristems after chalcone treatment, which reflects the important role of these enzymes on the mode of action of this secondary metabolite.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11970
1,3-Diphenyl-2-propenone, ≥98.0% (GC)
C15H12O