Clinical endocrinology
Single-nucleotide variants in two Hedgehog genes, SHH and HHIP, as genetic cause of combined pituitary hormone deficiency.
PMID 22897141
Combined pituitary hormone deficiency (CPHD) is characterized by deficiencies of two or more anterior pituitary hormones. Its genetic cause is unknown in the majority of cases. The Hedgehog (Hh) signalling pathway has been implicated in disorders associated with pituitary development. Mutations in Sonic Hedgehog (SHH) have been described in patients with holoprosencephaly (with or without pituitary involvement). Hedgehog interacting protein (HHIP) has been associated with variations in adult height in genome wide association studies. We investigated whether mutations in these two genes of the Hh pathway, SHH and HHIP, could result in 'idiopathic' CPHD. We directly sequenced the coding regions and exon - intron boundaries of SHH and HHIP in 93 CPHD patients of the Dutch HYPOPIT study in whom mutations in the classical CPHD genes PROP1, POU1F1, HESX1, LHX3 and LHX4 had been ruled out. We compared the expression of Hh genes in Hep3B transfected cells between wild-type proteins and mutants. We identified three single-nucleotide variants (p.Ala226Thr, c.1078C>T and c.*8G>T) in SHH. The function of the latter was severely affected in our in vitro assay. In HHIP, we detected a new activating variant c.-1G>C, which increases HHIP's inhibiting function on the Hh pathway. Our results suggest involvement of the Hedgehog pathway in CPHD. We suggest that both SHH and HHIP are investigated as a second screening in CPHD, after mutations in the classical CPHD genes have been ruled out.