EMAIL THIS PAGE TO A FRIEND
Contraception
Pharmacokinetic profile of nomegestrol acetate and 17β-estradiol after multiple and single dosing in healthy women.
PMID 22898360
Abstract
The pharmacokinetics of the monophasic oral contraceptive nomegestrol acetate (NOMAC) plus 17β-estradiol (E(2)) were investigated after a single dose and multiple dosing. NOMAC/E2 (2.5 mg/1.5 mg) was administered daily to healthy women (18-50 years, n=23) for 24 days; blood samples for pharmacokinetic analysis were obtained on Day 24 and again, after a 10-day pill-free interval, on Day 35 after a single dose. NOMAC reached steady state after 5 days with mean ±standard deviation (SD) trough NOMAC concentration (C(av)) of 4.4±1.4 ng/mL. On Day 24, mean±SD peak NOMAC concentration (Cmax, 12.3±3.5 ng/mL) was reached in mean 1.5 h (t(max)); the mean±SD elimination half-life (t(½)) was 45.9±15.3 h. After a single dose, NOMAC mean±SD C(max) was 7.2±2.0 ng/mL and mean±SD t(½) was 41.9±16.2 h. On Day 24, E2 mean±SD C(av) was 50.3±25.7 pg/mL; mean±SD Cmax was 86.0±51.3 pg/mL. After a single dose, mean±SD E2 Cmax was 253±179 pg/mL. These data demonstrate that NOMAC/E2 has a pharmacokinetic profile consistent with once-daily dosing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0133
Nomegestrol acetate, ≥98% (HPLC)
C23H30O4