EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
CoOOH nanosheet electrodes: Simple fabrication for sensitive electrochemical sensing of hydrogen peroxide and hydrazine.
PMID 22898661
Abstract
Cobalt oxyhydroxide, CoOOH, nanosheets were prepared via a surface alkaline treatment of cobalt foil at room temperature without using templates and catalysts. The morphology, chemical composition and structures of the nanosheets were characterized by XRD, FTIR and Raman spectroscopy, FESEM and TEM. These oriented and nanostructured arrays can be used directly as electrodes, thus simplifying the electrode fabrication process, as well as offering advantages such as enhanced electrode-electrolyte contact area, minimum diffusion resistance and direct active material-current collector connection for fast electron transport. The electrode was used as an electrochemical sensor towards non-enzymatic detection of hydrogen peroxide and hydrazine in alkaline solution. The amperometric detection of H(2)O(2) and N(2)H(4) was carried out at low potential (0V and 0.1V). At 0.1V, the amperometric signals are linearly proportional to H(2)O(2) concentration up to 1.6mM (R(2)=0.995), showing a detection limit (S/N=3) of 40μM and a high sensitivity of 99μA mM(-1)cm(-2). For N(2)H(4), the amperometric signals are linearly proportional to concentration up to 1.2mM (R(2)=0.99), showing a detection limit (S/N=3) of 20μM and a high sensitivity of 155μA mM(-1)cm(-2) at 0.1V.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF94594450
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
Co
GF41541476
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Co
GF45372315
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Co
GF08560289
Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%
Co
GF89559343
Cobalt, foil, 4mm disks, thickness 0.005mm, 99.9%
Co
GF48392051
Cobalt, foil, 4mm disks, thickness 0.006mm, 99.9%
Co
GF55954873
Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Co
GF92926727
Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%
Co
GF70445246
Cobalt, foil, 4mm disks, thickness 0.009mm, 99.9%
Co
GF40089225
Cobalt, foil, 4mm disks, thickness 0.0125mm, 99.9%
Co
GF48813723
Cobalt, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Co
GF10782121
Cobalt, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Co
GF40808516
Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.9%
Co
GF18093461
Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Co
GF30160291
Cobalt, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF81966224
Cobalt, foil, 100x100mm, thickness 0.7mm, annealed, 99.9%
Co
GF56494187
Cobalt, foil, 100x100mm, thickness 1.0mm, as rolled, 99.9%
Co
GF76259593
Cobalt, foil, 100x100mm, thickness 2.0mm, as rolled, 99.9%
Co
GF91564904
Cobalt, foil, 250x250mm, thickness 0.50mm, annealed, 99.9%
Co
GF88288699
Cobalt, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Co
GF30959411
Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Co
GF67617153
Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Co
GF85563535
Cobalt, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF09035647
Cobalt, foil, 25x25mm, thickness 0.7mm, annealed, 99.9%
Co
GF41834011
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.9%
Co
GF34805268
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Co
GF40133472
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.9%
Co
GF03773640
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.9%
Co
GF06754406
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Co
GF42159315
Cobalt, foil, light tested, 25x25mm, thickness 0.009mm, 99.9%
Co