EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Pseudonocardia hispaniensis sp. nov., a novel actinomycete isolated from industrial wastewater activated sludge.
PMID 22899021
Abstract
A novel actinomycete, designated PA3(T), was isolated from an oil refinery wastewater treatment plant, located in Palos de la Frontera, Huelva, Spain, and characterized taxonomically by using a polyphasic approach. Phylogenetic analysis based on 16S rRNA gene sequences showed that the isolate formed a distinct subclade in the Pseudonocardia tree together with Pseudonocardia asaccharolytica DSM 44247(T). The chemotaxonomic properties of the isolate, for example, the presence of MK-8 (H(4)) as the predominant menaquinone and iso-C(16:0) as the major fatty acid, are consistent with its classification in the genus Pseudonocardia. DNA:DNA pairing experiments between the isolate and the type strain of P. asaccharolytica DSM 44247(T) showed that they belonged to separate genomic species. The two strains were readily distinguished using a combination of phenotypic properties. Consequently, it is proposed that isolate PA3(T) represents a novel species for which the name Pseudonocardia hispaniensis sp. nov. is proposed. The type strain is PA3(T) (= CCM 8391(T) = CECT 8030(T)).
Related Materials