EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Acene-doped polymer films: singlet oxygen dosimetry and protein sensing.
PMID 22899174
Abstract
This paper describes thin films comprising acenes dispersed in a conjugated polymeric host that have a ratiometric photoluminescence response to singlet oxygen. These films also respond to irradiation of protein-bound sensitizers, which represents a solution to the problem of protein-conjugated polymer non-specific interactions.
Related Materials