EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Antioxidation and DNA-binding properties of binuclear lanthanide(III) complexes with a Schiff base ligand derived from 8-hydroxyquinoline-7-carboxaldehyde and benzoylhydrazine.
PMID 22899614
Abstract
8-Hydroxyquinoline-7-carboxaldehyde (8-HQ-7-CA), Schiff-base ligand 8-hydroxyquinoline-7-carboxaldehyde benzoylhydrazone, and binuclear complexes [LnL(NO(3))(H(2)O)(2)](2) were prepared from the ligand and equivalent molar amounts of Ln(NO(3))·6H(2)O (Ln=La(3+), Nd(3+), Sm(3+), Eu(3+), Gd(3+), Dy(3+), Ho(3+), Er(3+), Yb(3+), resp.). Ligand acts as dibasic tetradentates, binding to Ln(III) through the phenolate O-atom, N-atom of quinolinato unit, and C=N and -O-C=N- groups of the benzoylhydrazine side chain. Dimerization of this monomeric unit occurs through the phenolate O-atoms leading to a central four-membered (LnO)(2) ring. Ligand and all of the Ln(III) complexes can strongly bind to CT-DNA through intercalation with the binding constants at 10(5)-10(6) M(-1). Moreover, ligand and all of the Ln(III) complexes have strong abilities of scavenging effects for hydroxyl (HO·) radicals. Both the antioxidation and DNA-binding properties of Ln(III) complexes are much better than that of ligand.