EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of optically active 4-substituted 2-cyclohexenones.
PMID 22900684
Abstract
Recently, Nicolaou and Baran independently synthesized optically active 4-substituted 2-cyclohexenones via an efficient LiOH-mediated intramolecular aldol condensation. Thus far, application of their cyclization approach has been limited to ketoaldehydes where the R-group is branched. It is demonstrated that the LiOH-mediated cyclization, when applied to substrates containing unbranched R-groups, results in significant erosion of optical purity. A mechanistic justification is provided, and a set of neutral, organocatalyzed conditions is identified that enables cyclization with little loss in optical purity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C102814
2-Cyclohexen-1-one, ≥95%
C6H8O