EMAIL THIS PAGE TO A FRIEND
The American journal of medicine
Hypertension in older adults: progress and limitations.
PMID 22902034
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7106
Azelnidipine, ≥98% (HPLC), powder
C33H34N4O6