EMAIL THIS PAGE TO A FRIEND
Neoplasia (New York, N.Y.)
Sabutoclax, a Mcl-1 antagonist, inhibits tumorigenesis in transgenic mouse and human xenograft models of prostate cancer.
PMID 22904682
Abstract
Resistance to available therapeutic agents has been a common problem thwarting progress in treatment of castrate-resistant and metastatic prostate cancer (PCa). Overexpression of the Bcl-2 family members, including Mcl-1, in PCa cells is known to inhibit intracellular mitochondrial-dependent apoptosis. Here we report the development of a novel transgenic mouse model that spontaneously develops prostatic intraepithelial neoplasia and adenocarcinoma by the inducible, conditional knockout of transforming growth factor β receptor type II in stromal fibroblastic cells (Tgfbr2(ColTKO)). The Tgfbr2(ColTKO) prostate epithelia demonstrated down-regulation of luminal and basal differentiation markers, as well as Pten expression and up-regulation of Mcl-1. However, unlike in men, Tgfbr2(ColTKO) prostates exhibited no regression acutely after castration. The administration of Sabutoclax (BI-97C1), a pan-active Bcl-2 protein family antagonist mediated apoptosis in castrate-resistant PCa cells of Tgfbr2(ColTKO) mice and human subcutaneous, orthotopic, and intratibial xenograft PCa models. Interestingly, Sabutoclax had little apoptotic effect on benign prostate tissue in Tgfbr2(ColTKO) and wild-type mice. Sabutoclax was able to block c-Met activation, a critical axis in PCa metastatic progression. Further, Sabutoclax synergistically sensitized PC-3 cells to the cytotoxic effects of docetaxel (Taxotere). Together, these data suggest that Sabutoclax inhibits castrate-resistant PCa alone at the primary and bone metastatic site as well as support sensitivity to docetaxel treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8761
(±)-Gossypol from cotton seeds, ≥95% (HPLC)
C30H30O8
SML0433 AT101, ≥98% (HPLC)
C30H30O8