PloS one
Cationic amphiphilic drugs are potent inhibitors of yeast sporulation.
PMID 22905177
Meiosis is a highly regulated developmental process that occurs in all eukaryotes that engage in sexual reproduction. Previous epidemiological work shows that male and female infertility is rising and environmental factors, including pollutants such as organic solvents, are thought to play a role in this phenomenon. To better understand how organic compounds interfere with meiotic development, the model organism Saccharomyces cerevisiae was exposed to 446 bioactive molecules while undergoing meiotic development, and sporulation efficiency was quantified employing two different high-throughput assays. 12 chemicals were identified that strongly inhibited spore formation but did not interfere with vegetative growth. Many of these chemicals are known to bind to monoamine-receptors in higher eukaryotes and are cationic amphiphilic drugs. A detailed analysis of one of these drugs, tripelennamine, revealed that it induces sporulation-specific cytotoxicity and a strong inhibition of meiotic M phase. The drug, however, only mildly interfered with pre-meiotic DNA synthesis and the early meiotic transcriptional program. Chemical-genomic screening identified genes involved in autophagy as hypersensitive to tripelennamine. In addition, we found that growing and sporulating yeast cells heterozygous for the aminophospholipid translocase, NEO1, are haploinsufficient in the presence of the drug.
Related Materials
Product #
Molecular Formula
Add to Cart
Tripelennamine hydrochloride, analytical standard
C16H21N3 · HCl