EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Threshold voltage fluctuation in 16-nm-gate FinFets induced by random work function of nanosized metal grain.
PMID 22905489
Abstract
The random work-function (WK) induced threshold voltage fluctuation (sigmaVth) in 16 nm Titanium Nitride (TiN) metal-gate fin-type field effect transistors (FinFETs) is explored and modeled by using an experimentally validated Monte Carlo simulation approach. The influences of metal-grain size and device geometry aspect ratio on the random WK-induced sigmaVth are considered in the proposed equation analytically. The formula accounts for the inside of fluctuation and could be used for the assessment of effectiveness of suppression techniques.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
595063
Titanium nitride, <3 μm
NTi