EMAIL THIS PAGE TO A FRIEND
The Journal of international medical research
Effect of β-(3,4-dihydroxyphenyl)lactic acid on oxidative stress stimulated by high glucose levels in human peritoneal mesothelial cells.
PMID 22906267
Abstract
To investigate the effects of β-(3,4-dihydroxyphenyl)lactic acid on oxidative stress stimulated by high glucose levels in human peritoneal mesothelial cells (HPMCs) in vitro. HPMCs were incubated with 100 mol/l glucose followed by 0.625-20 mg/ml β-(3,4-dihydroxyphenyl)lactic acid. Reactive oxygen species (ROS) were quantified by flow cytometry. Relative levels of fibronectin-1 (FN1), collagen-I α(1) (COL1A1), endothelin-1 (EDN1) and haem oxygenase-1 (HMOX1) mRNA and protein were quantified by real-time reverse transcription-polymerase chain reaction and Western blotting, respectively. Absolute levels of FN1 and COLIA1 were quantified by enzyme-linked immunosorbent assay. β-(3,4-Dihydroxyphenyl)lactic acid significantly decreased ROS levels, and EDN1 mRNA and protein levels, in dose- and time-dependent manners. HMOX1 mRNA and protein levels were significantly increased by β-(3,4-dihydroxyphenyl)lactic acid in dose-dependent manners. COL1A1 and FN1 mRNA and protein levels were significantly decreased by β-(3,4-dihydroxyphenyl)lactic acid in dose- and time-dependent manners. β-(3,4-Dihydroxyphenyl)lactic acid inhibited oxidative stress and reversed increases in FN1 and COLIA1 induced by high glucose levels in HPMCs. This may contribute to a protective role in peritoneal fibrosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0679
Danshensu sodium salt, ≥98% (HPLC)
C9H9O5 · Na