EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Alkaloids isolated from the lateral root of Aconitum carmichaelii.
PMID 22907155
Abstract
Two new alkaloids, aconicarmine (1) and aconicaramide (5), were isolated from the EtOH extract of the lateral roots of Aconitum carmichaelii, together with five known compounds: fuziline (2), neoline (3), N-ethylhokbusine B (4), 5-hydroxymethylpyrrole-2-carbaldehyde (6), and oleracein E (7). Their structures were elucidated by physical and NMR analysis. Pyrrole alkaloids were isolated from A. carmichaelii for the first time. In the in vitro assays, compounds 2 and 3 showed activity against pentobarbital sodiuminduced cardiomyocytes damage by obviously recovering beating rhythm and increasing the cell viability, while compounds 5 and 7 showed moderate antibacterial activity.